Mouse nerve growth factor 7S (from submaxillary glands)
Lieferant: ENZO LIFE SCIENCES
ALX521008C100
ENZOALX521008C100EA 480 EUR
ENZOALX521008C100
Mouse nerve growth factor 7S (from submaxillary glands)
Proteine und Peptide
Isolated from mouse submaxillary glands by a modification of the procedure described by T. Darling, et al.; Cell Culture Meth. Mol. Cell Biol. 4, 79 (1984).
Quantity: Determined by Lowry and Pierce method.