Human recombinant Fas:Fc
Lieferant: ENZO LIFE SCIENCES
ALX522002C050
ENZOALX522002C050EA 550 EUR
ENZOALX522002C050
Human recombinant Fas:Fc
Proteine und Peptide
Produced in HEK 293 cells. The extracellular domain of human Fas (CD95; APO-1) (aa 7-154) is fused to the Fc portion of human IgG1.