Human recombinant CD40:Fc
Lieferant: ENZO LIFE SCIENCES
ALX522016C050
ENZOALX522016C050EA
550
EUR
ENZOALX522016C050
Human recombinant CD40:Fc
Proteine und Peptide
Produced in HEK 293 cells. The cysteine-rich region (aa 21-193) of human CD40 is fused to the Fc portion of human IgG1.