Human recombinant LIGHT (soluble)
Lieferant: ENZO LIFE SCIENCES
ALX522018C010
ENZOALX522018C010EA 645 EUR
ENZOALX522018C010
Human recombinant LIGHT (soluble)
Proteine und Peptide
Produced in HEK 293 cells. The extracellular domain of human LIGHT (aa 89-240) is fused at the N-terminus to a linker peptide (10 aa) and a FLAG®-tag.