Human recombinant nerve growth factor receptor:Fc
Lieferant: ENZO LIFE SCIENCES
ALX522044C050
ENZOALX522044C050EA 591 EUR
ENZOALX522044C050
Human recombinant nerve growth factor receptor:Fc
Proteine und Peptide
Produced in HEK 293 cells. The cysteine-rich domain of human NGFR (nerve growth factor receptor; p75NTR) (aa 1-250) is fused to the Fc portion of human IgG1.