Human recombinant TL1A (soluble) (from E. coli)
Lieferant: ENZO LIFE SCIENCES
ALX522084C010
ENZOALX522084C010EA 624 EUR
ENZOALX522084C010
Human recombinant TL1A (soluble) (from E. coli)
Proteine und Peptide
Produced in E. coli. The extracellular domain of human TL1A (aa 93-251) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.