Human recombinant LRRC32:Fc (from CHO cells)
Lieferant: ENZO LIFE SCIENCES
ALX522117C050
ENZOALX522117C050EA 599 EUR
ENZOALX522117C050
Human recombinant LRRC32:Fc (from CHO cells)
Proteine und Peptide
Produced in CHO cells. Human LRRC32 (aa 20-627) is fused to the Fc portion of human IgG1.