Mouse recombinant netrin 1
Lieferant: ENZO LIFE SCIENCES
ALX5221242010
ENZOALX5221242010EA 673 EUR
ENZOALX5221242010
Mouse recombinant netrin 1
Proteine und Peptide
Produced in HEK 293 cells. The extracellular domain of mouse netrin-1 (aa 27-604) is fused at the C-terminus to a FLAG®-tag.