(+/-)-2-Amino-5-phosphonopentanoic acid ≥99% (durch HPLC, TLC)
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
Synonyme:
DL-2-amino-5-phosphonovaleric acid, DL-AP5, 2-Amino-5-phosphonopentanoic-acid
ALX550034M025
ENZOALX550034M025EA
433
EUR
ENZOALX550034M025
(+/-)-2-Amino-5-phosphonopentanoic acid ≥99% (durch HPLC, TLC)
(+/-)-2-Amino-5-phosphonopentanoic acid
NMDA receptor antagonist.
Potent and selective NMDA receptor antagonist.
Soluble in 100 mM NaOH or 25 mM water.
Formel:
C₅H₁₂NO₅P Molecular Weight: 197,127 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
76326-31-3 |
Spezifikation Testergebnisse
Purity (HPLC, TLC) | ≥99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...