6-Cyan-7-nitrochinoxalin-2,3-dion ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

ALX550042M005 ALX550042M010 ALX550042M025 ALX550042M050
ENZOALX550042M005EA 85.8 EUR
ENZOALX550042M005 ENZOALX550042M010 ENZOALX550042M025 ENZOALX550042M050
6-Cyan-7-nitrochinoxalin-2,3-dion ≥98% (durch TLC)
6-Cyan-7-nitrochinoxalin-2,3-dion

AMPA/kainate receptor antagonist


  • Formulation: Off-white to yellow powder.


Potent, competitive kainate/quisqualate (non-NMDA) receptor antagonist. For the more soluble form see CNQX . disodium salt (Prod. No. ALX-550-283).

Formel: C₉H₄N₄O₄
MDL: MFCD00069232
CAS-Nummer: 115066-14-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR