6-Cyan-7-nitrochinoxalin-2,3-dion ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
ALX550042M005
ALX550042M010
ALX550042M025
ALX550042M050
ENZOALX550042M005EA
85.8
EUR
ENZOALX550042M005
ENZOALX550042M010
ENZOALX550042M025
ENZOALX550042M050
6-Cyan-7-nitrochinoxalin-2,3-dion ≥98% (durch TLC)
6-Cyan-7-nitrochinoxalin-2,3-dion
AMPA/kainate receptor antagonist
- Formulation: Off-white to yellow powder.
Potent, competitive kainate/quisqualate (non-NMDA) receptor antagonist. For the more soluble form see CNQX . disodium salt (Prod. No. ALX-550-283).
Formel:
C₉H₄N₄O₄ |
MDL:
MFCD00069232 CAS-Nummer: 115066-14-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...