R-(+)-Baclofen hydrochloride ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
R-(+)-β-(Aminomethyl)-4-chlorobenzenepropanoic acid.HCl
ALX550074M050
ENZOALX550074M050EA
198
EUR
ENZOALX550074M050
R-(+)-Baclofen hydrochloride ≥98% (durch HPLC)
R-(+)-Baclofen hydrochloride
Selective GABAB receptor agonist.
Skeletal muscle relaxant. The more active enantiomer of (±)-baclofen.
Soluble in water, dilute aqueous base or DMSO.
Formel:
C₁₀H₁₂ClNO₂·HCl Molecular Weight: 250,2 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
63701-55-3 Merck Index: 14,00937 |
Spezifikation Testergebnisse
Purity (HPLC) | ≥98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...