L-Noradrenaline bitartrate monohydrate ≥98%

Lieferant: ENZO LIFE SCIENCES
Gefahr

Synonyme: L-Arterenol Bitartrate, L-Norepinephrine Bitartrate

ALX550076G001
ENZOALX550076G001EA 260 EUR
ENZOALX550076G001
L-Noradrenaline bitartrate monohydrate ≥98%
L-Noradrenaline bitartrate monohydrate

Adrenergic neurotransmitter.


Soluble in water (46 mg/ml).

Formel: C₈H₁₁NO₃
Molecular Weight: 319,27 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00036384
CAS-Nummer: 108341-18-0
UN: 2811
ADR: 6.1,II
Merck Index: 14,06695

Bestellen

Spezifikation Testergebnisse

Purity (assay) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR