AIDA ≥96%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
(±)-1-Aminoindane-1,5-dicarboxylic acid
ALX550101M025
ENZOALX550101M025EA
260
EUR
ENZOALX550101M025
AIDA ≥96%
AIDA
mGluR agonist.
Potent and selective ligand for group I mGluRs. Shows no effect on group II or group III mGluRs. Has no effect on ionotropic glutamate receptors.
Soluble in water.
Formel:
C₁₁H₁₁NO₄ Molecular Weight: 221,2 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
168560-79-0 |
Spezifikation Testergebnisse
Purity | ≥96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...