D-(+)-Neopterin ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
Synonyme:
2-Amino-6-((1S,2R)-1,2,3-trihydroxypropyl)pteridin-4(3H)-one, Neopterin, D(+)-6-(erythro-1,2,3-Trihydroxypropyl)pterin
ALX550126M050
ENZOALX550126M050EA
158
EUR
ENZOALX550126M050
D-(+)-Neopterin ≥98% (durch HPLC)
D-(+)-Neopterin
Biochemical marker used as an indicator of cell proliferation. Pterin cofactor. Precursor of tetrahydrobiopterin.
Soluble in 0.05 M NaOH or 1M HCl; very slightly soluble in water. Ultrasonication may improve the dissolution.
Formel:
C₉H₁₁N₅O₄ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00042801 CAS-Nummer: 2009-64-5 EINECS: 217-924-7 Merck Index: 14,06462 |
Spezifikation Testergebnisse
Purity (HPLC) | ≥98% |
Appearance | Off-white to yellow solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...