Histamin Dihydrochlorid
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Histamine 2HCl, 4-(2-Aminoethyl)-1H-imidazole dihydrochloride, 2-(1H-Imidazol-4-yl)ethan-1-amine dihydrochloride
ALX550132G005
ENZOALX550132G005EA
53.9
EUR
ENZOALX550132G005
Histamin Dihydrochlorid
Histamin Dihydrochlorid
Formel:
C₅H₁₁Cl₂N₃ Molecular Weight: 184,07 g/mol |
MDL:
MFCD00012703 CAS-Nummer: 56-92-8 EINECS: 200-298-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...