Benztropine mesylate ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: Cogentin

ALX550143G001
ENZOALX550143G001EA 70.4 EUR
ENZOALX550143G001
Benztropine mesylate ≥98%
Benztropine mesylate

Muscarinic acetylcholine receptor antagonist


Muscarinic acetylcholine receptor antagonist. Inhibits the dopamine transporter.

Formel: C₂₂H₂₉NO₄S
Molecular Weight: 403,537 g/mol
MDL: MFCD00074784
CAS-Nummer: 132-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR