Amiloride hydrochloride dihydrate ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: N-Amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride dihydrate, Modamide, Amiprazidine, Amiloride HCl dihydrate, N-Amidino-3,5-diamino-6-chloropyrazinecarboxamide Hydrochloride dihydrate, Amiloride hydrochloride dihydrate, 3,5-Diamino-N-(aminoiminomethyl)-6-chloropyrazinecarboxamide hydrochloride dihydrate, 3,5-Diamino-N-carbamimidoyl-6-chloropyrazine-2-carboxamide Hydrochloride dihydrate

ALX550212G001
ENZOALX550212G001EA 67.1 EUR
ENZOALX550212G001
Amiloride hydrochloride dihydrate ≥98%
Amiloride hydrochloride dihydrate

Ca²⁺ channel inhibitor


Selectively blocks low threshold (T-type) calcium channel (KD=30μM) with no effect on L-type channels or tetrodotoxin Na+ channels. Inhibits the epithelial tetrodotoxin insensitive Na+ channel (IC50 =1μM). Clinically useful K+ sparing diuretic.

Formel: C₆H₈ClN₇O·HCl·2H₂O
Molecular Weight: 302,116 g/mol
MDL: MFCD00058197
CAS-Nummer: 17440-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR