Lidocain Hydrochlorid

Lieferant: Enzo Life Sciences

ALX550251G005
ENZOALX550251G005EA 170 EUR
ENZOALX550251G005
Lidocain Hydrochlorid
Lidocain Hydrochlorid
Formel: C₁₄H₂₃ClN₂O
CAS-Nummer: 73-78-9
EINECS: 200-803-8
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR