5-(N-Ethyl-N-isopropyl)amiloride ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 3-Amino-N-carbamimidoyl-6-chloro-5-(ethyl(isopropyl)amino)pyrazine-2-carboxamide, EIPA

ALX550266M005
ENZOALX550266M005EA 85.8 EUR
ENZOALX550266M005
5-(N-Ethyl-N-isopropyl)amiloride ≥98% (durch TLC)
5-(N-Ethyl-N-isopropyl)amiloride

Selective inhibitor of the Na /H antiporter.


  • Na⁺/H⁺ antiporter inhibitor


Off-white to yellow solid.


Soluble in methanol, DMSO or dilute aqueous acid; insoluble in water.

Formel: C₁₁H₁₈ClN₇O
Molecular Weight: 299,763 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00151710
CAS-Nummer: 1154-25-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR