Nicardipin -hydrochlorid ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 3-(2-(Benzyl(methyl)amino)ethyl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate hydrochloride, Nicardipine HCl

ALX550273G005
ENZOALX550273G005EA 171 EUR
ENZOALX550273G005
Nicardipin -hydrochlorid ≥98%
Nicardipin -hydrochlorid

Ca²⁺ channel inhibitor


Ca²⁺ channel antagonist. Antihypertensive.

Formel: C₂₆H₂₉N₃O₆
Molecular Weight: 515,99 g/mol
MDL: MFCD00057327
CAS-Nummer: 54527-84-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR