Nimodipin

Lieferant: ENZO LIFE SCIENCES

Synonyme: 3-Isopropyl 5-(2-methoxyethyl) 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Isopropyl 2-Methoxyethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate

ALX550277G001
ENZOALX550277G001EA 340 EUR
ENZOALX550277G001
Nimodipin
Nimodipin

Ca²⁺ channel inhibitor


Ca²⁺ channel antagonist. Highly potent at L-type channels. Block some low-voltage activated Ca²⁺ channels. Enhances memory in old or brain-damaged animals. Ameliorates experimental diabetic neuropathy in streptozotocin-induced diabetic rats. Vasodilator.

Formel: C₂₁H₂₆N₂O₇
Molecular Weight: 418,45 g/mol
MDL: MFCD00153848
CAS-Nummer: 66085-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR