Praziquantel ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
2-(Cyclohexylcarbonyl)-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, 2-(Cyclohexanecarbonyl)-2,3,6,7-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4(11bH)-one
ALX550321G005
ENZOALX550321G005EA
50.6
EUR
ENZOALX550321G005
Praziquantel ≥98% (durch HPLC)
Praziquantel
Antihelmintic agent.
Soluble in 100% ethanol or chloroform; slightly soluble in water.
Formel:
C₁₉H₂₄N₂O₂ Molecular Weight: 312.41 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00058531 CAS-Nummer: 55268-74-1 Merck Index: 14,07716 |
Spezifikation Testergebnisse
Purity (HPLC) | ≥98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...