Galantamin Hydrobromid ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Galanthamine HBr, (4aS,6R,8aS)-3-Methoxy-11-methyl-5,6,9,10,11,12-hexahydro-4aH-benzo[2,3]benzofuro[4,3-cd]azepin-6-ol hydrobromide

ALX550336M050
ENZOALX550336M050EA 217 EUR
ENZOALX550336M050
Galantamin Hydrobromid ≥98% (durch HPLC)
Galantamine hydrobromide

Specific, competitive and reversible acetylcholinesterase inhibitor. Alzheimer's disease therapeutic.


  • Acetylcholine esterase inhibitor


White to off-white crystalline solid.


Soluble in DMSO (10 mg/ml) or 100% ethanol (4 mg/ml); sparingly soluble in water.

Formel: C₁₇H₂₁NO₃·HBr
Molecular Weight: 368,27 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00067672
CAS-Nummer: 1953-04-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR