3,5-Dimethyl-1-adamantanamine hydrochloride ≥98% (durch argentometrische Titration)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Memantine Hydrochloride, 1-Amino-3,5-dimethyladamantane Hydrochloride
ALX550363M050
ENZOALX550363M050EA
103
EUR
ENZOALX550363M050
3,5-Dimethyl-1-adamantanamine hydrochloride ≥98% (durch argentometrische Titration)
3,5-Dimethyl-1-adamantanamine hydrochloride
NMDA receptor antagonist.
Low-affinity NMDA receptor antagonist. Stimulates dopamine release.
Formel:
C₁₂H₂₁N Molecular Weight: 215.77 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00214336 CAS-Nummer: 41100-52-1 EINECS: 255-219-6 Merck Index: 14,05829 |
Spezifikation Testergebnisse
Purity (argentmetric titration) | ≥98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...