MSK-195 ≥98%, weißer Feststoff
Lieferant: ENZO LIFE SCIENCES
ALX550396M005
ENZOALX550396M005EA
534
EUR
ENZOALX550396M005
MSK-195 ≥98%, weißer Feststoff
MSK-195
TRP channel agonist.
Potent analgesic with EC₅₀=0.96 µg/kg in the acetic acid-induced writhing test. RTX binding affinity (Ki=603 nM), agonism (calcium influx; EC₅₀=240 nM).
Soluble in methanol, 100% ethanol, DMSO or other organic solvents; insoluble in water.
Formel:
C₂₈H₄₀N₂O₅ Molecular Weight: 484,6 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
289902-82-5 |
Spezifikation Testergebnisse
Purity | ≥98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...