Dimethyl-W84 dibromide ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

ALX550400M005
ENZOALX550400M005EA 192 EUR
ENZOALX550400M005
Dimethyl-W84 dibromide ≥98% (durch HPLC)
Dimethyl-W84 dibromide

M2 receptor modulator.


Very potent allosteric modulator of muscarinic receptors, selective for M2-receptors.


Retards the dissociation of the antagonist N-methylscopolamine. Affinity for the free receptor is about 10-fold higher than with W84. pKA=7.08, p(αKA)=6.75, pEC₅₀,diss=6.77.


Soluble in DMSO, water or aqueous buffer.

Formel: C₃₄H₄₈Br₂N₄O₄
Molecular Weight: 736,6 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 402475-33-6

Bestellen

Spezifikation Testergebnisse

Purity (HPLC) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR