L-Kynurenine ≥95% (durch HPCE)

Lieferant: ENZO LIFE SCIENCES

Synonyme: β-Anthraniloyl-L-alanine, (S)-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid, L-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid, Kynurenine

ALX550408M050
ENZOALX550408M050EA 50.6 EUR
ENZOALX550408M050
L-Kynurenine ≥95% (durch HPCE)
L-Kynurenine

Key intermediate in the breakdown pathway of tryptophan.


Soluble in 0.5 M hydrochloric acid (50 mg/ml); very poorly soluble in water (0.4 g/100 ml, 20 °C).

Formel: C₁₀H₁₂N₂O₃
Molecular Weight: 208,22 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00069912
CAS-Nummer: 2922-83-0
Merck Index: 14,05328

Bestellen

Spezifikation Testergebnisse

Purity (HPLC) ≥95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR