Lipopolysaccharides (aus E. coli) serotype EH100 (Ra), TLRgrade™ anwendungsbereit
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
TLR4 activator.
Rough (R)-form LPS isolated and purified from E. coli EH100 (Ra-mutant) by a modification of the PCP extraction method, converted to the uniform sodium salt form.
Strong activator of Toll-like receptor (TLR) 4. Does not activate TLR2 or other TLRs as determined with splenocytes and macrophages from TLR4 deficient mice. No further re-extraction required. Smooth (S)-form LPS are commonly the preferred choice for whole animal studies, whereas Rough (R)-form LPS are primarily used in cellular in vitro activation studies.
Spezifikation Testergebnisse
Concentration | 1 mg/ml |
Formulation | Liquid. Sterile, ready-to-use solution in double distilled, pyrogen-free water |
Purity | Absence of detectable protein or DNA contaminants with agonistic TLR activity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...