Lipopolysaccharides 1 mg/ml in Wasser (aus S. enteritidis) S-Form, TLRgrade™ anwendungsbereit

Lieferant: ENZO LIFE SCIENCES
Gefahr

ALX581019L002
ENZOALX581019L002EA 324 EUR
ENZOALX581019L002
Lipopolysaccharides 1 mg/ml in Wasser (aus S. enteritidis) S-Form, TLRgrade™ anwendungsbereit
Lipopolysaccharides

TLR4 activator.


Smooth (S)-form LPS isolated and purified from Salmonella enteritidis (wild type) by a modification of the PCP extraction method, converted to the uniform sodium salt form.


Specific activator of Toll-like receptor 4 (TLR4). Does not activate TLR2 or other TLRs as determined with splenocytes and macrophages from TLR4 deficient mice. Smooth (S)-form LPS are commonly the preferred choice for whole animal studies, whereas Rough (R)-form LPS are primarily used in cellular in vitro activation studies.

Bestellen

Spezifikation Testergebnisse

Concentration 1 mg/ml
Formulation Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
Purity Absence of detectable protein or DNA contaminants with agonistic TLR activity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR