Lipopolysaccharides (aus S. abortus equi) S-form, TLRgrade™ anwendungsbereit
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
TLR4 activator.
Smooth (S)-form LPS, isolated and purified from Salmonella abortus equi by modification of the phenol water extraction and PCP method. LPS-Biotin was prepared using the biotin reagent biotinamidocaproate N-hydroxysuccinimide ester.
Useful reagent for evaluating LPS binding and cell activation in white blood cells.
Spezifikation Testergebnisse
Concentration | 1 mg/ml |
Formulation | Liquid. Sterile, ready-to-use solution in pyrogen-free double-distilled water |
Purity | Absence of detectable protein or DNA contaminants with agonistic TLR activity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...