Monophosphoryl lipid A 0,5 mg/l in Wasser (synthetisch) anwendungsbereit
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Glucopyranosyl lipid adjuvant, GLA, MPLA, MPL-A
ALX581205C100
ENZOALX581205C100EA
373
EUR
ENZOALX581205C100
Monophosphoryl lipid A 0,5 mg/l in Wasser (synthetisch) anwendungsbereit
Monophosphoryl lipid A
TLR4 activator.
- Sterile solution in pyrogen free double distilled water.
Activates Toll-like receptor 4 (TLR4) about 10-fold less (100 ng/ml) compared to the endotoxic lipid A molecule. Synthetic lipid A does not activate TLR2 as determined with splenocytes and macrophages from TLR4 deficient mice. Exhibits reduced endotoxic and pyrogenic activity compared to lipid A and LPS.
Formel:
C₉₆H₁₈₄N₃O₂₂P Molecular Weight: 1763.47 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
1246298-63-4 |
Spezifikation Testergebnisse
Source | Synthetic |
Concentration | 0.5 mg/ml |
Formulation | Liquid. Sterile, ready-to-use solution in pyrogen free double distilled water. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...