IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: IPTG

ALX582012G005 ALX582012G025
ENZOALX582012G005EA 154 EUR
ENZOALX582012G005 ENZOALX582012G025
IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥98% (durch HPLC)
IPTG (Isopropyl-β-D-thiogalactopyranosid)

β-galactosidase activator.


Inducer of β-D-galactosidase. Used in the stimulation of β-galactosidase in cellular systems in which dioxane would disrupt normal cell function. Contains no animal-derived products.


Soluble in water (5%) or methanol.

Formel: C₉H₁₈O₅S
Molecular Weight: 238.31 g/mol
Siedepunkt: 402
Lagertemperatur: Gefrierschrank
MDL: MFCD00063273
CAS-Nummer: 367-93-1
EINECS: 206-703-0

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (HPLC)
Identity Determined by IR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR