DAF-2 ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: 2-​(3,​6-​dihydroxy-​4,​5-​diamino-​9H-​xanthen-​9-​yl)-​benzoic acid, 4,5-Diaminofluorescein

ALX620052M001
ENZOALX620052M001EA 608 EUR
ENZOALX620052M001
DAF-2 ≥98% (durch HPLC)
DAF-2

Fluorescent probe for NO detection.


Highly sensitive fluorescent probe for the real-time detection of nitric oxide (NO) in vivo. In the cell, the product reacts rapidly with NO in the presence of O₂ to form the highly fluorescent compound triazolofluorescein (DAF-2T, Ex 495 nm; Em 515 nm).

Formel: C₂₀H₁₆N₂O₅
Molecular Weight: 364,34 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD01866182
CAS-Nummer: 205391-01-1
EINECS: 200-664-3

Bestellen

Spezifikation Testergebnisse

Concentration ~5 mM
Formulation Dissolved in 0.55 ml DMSO
Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR