JC-1 iodide salt ≥98% (durch HPLC), ultrarein

Lieferant: ENZO LIFE SCIENCES

Synonyme: JC-1

ALX620053M005
ENZOALX620053M005EA 358 EUR
ENZOALX620053M005
JC-1 iodide salt ≥98% (durch HPLC), ultrarein
JC-1 iodide salt

Fluorescent probe to test Pgp activity


JC-1 is a very sensitive fluorescent probe to test Pgp activity. In acute myeloid leukemia (AML) samples JC-1 is capable of distinguishing between resistant, intermediate and sensitive groups, whereas rhodamine 123 only recognizes the resistant group. As in cell lines, the red emission band (597nm) of JC-1 appears to be more convenient for detection of low-level resistance in AML than other probes, such as rhodamine 123 (529nm) or calcein-AM (517nm). Green fluorescent JC-1 existing as a monomer (

Formel: C₂₅H₂₇Cl₄IN₄
Molecular Weight: 652,23 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00188475
CAS-Nummer: 47729-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR