DAF-2 diacetate 5 mM in DMSO ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: 2-​(3,​6-​diacetyloxy-​4,​5-​diamino-​9H-​xanthen-​9-​yl)-​benzoic acid, DAF-2 DA, 4,5-Diaminofluorescein diacetate

ALX620056M001
ENZOALX620056M001EA 669 EUR
ENZOALX620056M001
DAF-2 diacetate 5 mM in DMSO ≥98% (durch HPLC)
DAF-2 diacetate

Fluorescent probe for NO detection. Cell permeable.


  • FORMULATION: 1 mg dissolved in 0.45ml DMSO.


Cell permeable analog of DAF-2 that is hydrolysed to DAF-2 by intracellular esterases. Can be used in fluorescence microscopy to measure real-time changes in nitric oxide (NO) levels in vivo (Ex(max): 495 nm; Em(max): 515 nm).

Formel: C₂₄H₁₈N₂O₇
Molecular Weight: 446,41 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD05865101
CAS-Nummer: 205391-02-2
EINECS: 200-664-3

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (HPLC)
Formulation Dissolved in 0.45 ml DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR