DAF-FM diacetate 2,5 mg/ml ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: DAF-FM DA, 4-Amino-5-methylamino-2',7'-difluorofluorescein diacetate

ALX620071M001
ENZOALX620071M001EA 1060 EUR
ENZOALX620071M001
DAF-FM diacetate 2,5 mg/ml ≥98% (durch HPLC)
DAF-FM diacetate

Fluorescent probe for NO detection. Cell permeable.


Cell permeable photo-stable nitric oxide (NO) fluorescent indicator, that is hydrolysed to DAF-FM by intracellular esterases (Ex: 500 nm; Em: 515 nm). Stable in a pH range of 5.5 – 12. High yield (quantum efficiency). Detection limit 3 nM.

Formel: C₂₅H₁₈F₂N₂O₇
Molecular Weight: 496,4 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 254109-22-3
EINECS: 200-664-3

Bestellen

Spezifikation Testergebnisse

Formulation Dissolved in 0.4 ml DMSO
Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR