HPF ~5mM ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Gefahr

Synonyme: Hydroxyphenyl fluorescein

ALX620074M001
ENZOALX620074M001EA 776 EUR
ENZOALX620074M001
HPF ~5mM ≥98% (durch HPLC)
HPF

Fluorescent probe for hROS detection.


Cell permeable fluorescent reagent (Ex(max): 490 nm; Em(max): 515 nm) for the detection of highly reactive oxygen species (hROS). Immediately reacts with hROS such as hydroxyl radical and peroxynitrite, and the fluorescence intensity greatly increases. In addition, peroxynitrite can be detected in distinction from nitric oxide and superoxide since HPF does not react with nitric oxide, superoxide and hydrogen peroxide.


HPF is resistant to light-induced autooxidation. HPF does not react with hypochlorite (-OCl) either and thus can be used in combination with APF which detects -OCl to elucidate reliably the roles of -OCl in biological systems such as neutrophils. Moreover, HPF is resistant to light-induced autooxidation.

Formel: C₂₆H₁₆O₆
Molecular Weight: 424,4 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 359010-69-8
EINECS: 200-679-5
UN: 2265
ADR: 3,III

Bestellen

Spezifikation Testergebnisse

Concentration ~5 mM
Formulation Dissolved in 0.47 ml dimethyl formamide
Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR