ZnAF-2 DA
Lieferant: ENZO LIFE SCIENCES
ALX620076M001
ALX620076M001
ALEXALX620076M001EA
637
EUR
ALEXALX620076M001
ENZOALX620076M001
ZnAF-2 DA
Proteinkonjugate und Markierungen
Fluorescent probe for Zn²⁺. Cell permeable.
Cell permeable derivative of ZnAF-2 (Prod. No. ALX-620-072). Fluorescent reagent (Ex(max): 492 nm; Em(max): 514 nm) for the detection of low concentration of zinc ion due to its strong affinity to zinc ion (dissociation constant: 2,7 nM). The sample zinc ion can be specifically detected. Low background fluorescense supersensitizes the visualization for in vivo sample zinc ion.