Aflatoxin M1 ≥95% (durch HPLC, TLC)
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
ALX630095MC01
ENZOALX630095MC01EA
162
EUR
ENZOALX630095MC01
Aflatoxin M1 ≥95% (durch HPLC, TLC)
Aflatoxin M1
Hepatotoxic and hepatocarcinogenic mycotoxin.
Potent hepatotoxic and hepatocarcinogenic mycotoxin. Hydroxylated metabolite of aflatoxin B1. Maximum absorbance (λmax) at 228, 264 and 360 nm.
Soluble in methylene chloride (1 mg/ml).
Formel:
C₁₇H₁₂O₇ Lagertemperatur: Kühlschrank |
CAS-Nummer:
6795-23-9 EINECS: 229-865-4 UN: 2811 ADR: 6.1,I Merck Index: 14,00182 |
Spezifikation Testergebnisse
Source | Isolated from Aspergillus flavus |
Purity | ≥95% (HPLC, TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...