Zearalenone ≥99% (durch HPLC, TLC)

Lieferant: ENZO LIFE SCIENCES
Gefahr

Synonyme: Benzoxacyclotetradecin-1,7(8H)-dione, (S,E)-14,16-Dihydroxy-3-methyl-3,4,5,6,9,10-hexahydro-1H-benzo[c][1]oxacyclotetradecine-1,7(8H)-dione

ALX630105M010
ENZOALX630105M010EA 175 EUR
ENZOALX630105M010
Zearalenone ≥99% (durch HPLC, TLC)
Zearalenone

Mycotoxin.


Zearalenone (ZEN) is an estrogenic mycotoxin in animals and a phytohormone in plants. Inducer of sister chromatid exchange and chromosomal aberration. Acts as a protonophoric uncoupler in plant mitochondria.


Soluble in methanol (5 mg/ml).

Formel: C₁₈H₂₂O₅
Molecular Weight: 318,4 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 17924-92-4
EINECS: 241-864-0
UN: 2811
ADR: 6.1,II
Merck Index: 14,10116

Bestellen

Spezifikation Testergebnisse

Source Isolated from Giberella zeae
Purity ≥99% (HPLC, TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR