Aflatoxin G2 ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
ALX630106M001
ALX630106M005
ENZOALX630106M001EA
406
EUR
ENZOALX630106M001
ENZOALX630106M005
Aflatoxin G2 ≥98% (durch TLC)
Aflatoxin G2
Hepatotoxic and hepatocarcinogenic mycotoxin.
Soluble in methylene chloride (5 mg/ml), DMSO or methanol.
Formel:
C₁₇H₁₄O₇ Molecular Weight: 330.3 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
7241-98-7 EINECS: 230-643-4 UN: 3462 ADR: 6.1 I Merck Index: 14,00181 |
Spezifikation Testergebnisse
Source | Isolated from Aspergillus flavus |
Purity | ≥98% (TLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...