Aflatoxin G2 ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES
Gefahr

ALX630106M001 ALX630106M005
ENZOALX630106M001EA 406 EUR
ENZOALX630106M001 ENZOALX630106M005
Aflatoxin G2 ≥98% (durch TLC)
Aflatoxin G2

Hepatotoxic and hepatocarcinogenic mycotoxin.


Soluble in methylene chloride (5 mg/ml), DMSO or methanol.

Formel: C₁₇H₁₄O₇
Molecular Weight: 330.3 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 7241-98-7
EINECS: 230-643-4
UN: 3462
ADR: 6.1 I
Merck Index: 14,00181

Bestellen

Spezifikation Testergebnisse

Source Isolated from Aspergillus flavus
Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR