Aflatoxin M2 ≥98% (durch HPLC, TLC)
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
Synonyme:
4-Hydroxyaflatoxin B2
ALX630114MC01
ENZOALX630114MC01EA
621
EUR
ENZOALX630114MC01
Aflatoxin M2 ≥98% (durch HPLC, TLC)
Aflatoxin M2
Hepatotoxic and hepatocarcinogenic mycotoxin.
Potent hepatotoxic and hepatocarcinogenic mycotoxin. Metabolite of aflatoxin B₂. DNA-damaging agent.
Soluble in DMSO or methanol, insoluble in water.
Formel:
C₁₇H₁₄O₇ Lagertemperatur: Gefrierschrank |
CAS-Nummer:
6885-57-0 EINECS: 229-989-9 UN: 3462 ADR: 6.1,I Merck Index: 14,00182 |
Spezifikation Testergebnisse
Source | Isolated from Aspergillus flavus |
Purity | ≥98% (HPLC, TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...