Astrovirusen
Lieferant: Enzo Life Sciences
ALX6301220200
ENZOALX6301220200EA
916
EUR
ENZOALX6301220200
Astrovirusen
Proteine und Peptide
Concentrated source of Astrovirus particles and proteins.
From astrovirus type 1, strain A88/2 Newcastle. Inactivated by b-propiolactone 0.1% and 4 hours at 32°C.