ODN 1668 (synthetisch), TLRgrade™, steril, gefriergetrocknet

Lieferant: ENZO LIFE SCIENCES

ALX746001C100
ENZOALX746001C100EA 234 EUR
ENZOALX746001C100
ODN 1668 (synthetisch), TLRgrade™, steril, gefriergetrocknet
ODN 1668

TLR9 ligand.


CpG oligodeoxynucleotide (Type B) with phosphorothioate backbone. Specific ligand for mouse TLR9 (Toll-like receptor 9).


Sufficient for at least 100 cellular activation assays at 0.1 – 0.5 µg/well when performed in a 200 µl assay volume (i.e. 96 well microtiter plate).


Lieferumfang: Includes 1 vial of double distilled water (endotoxin-free).

Bestellen

Spezifikation Testergebnisse

Source Synthetic
Purity  Activity and endotoxin tested - TLRgrade™
Endotoxin Content <0.002 EU/µg
Formulation Lyophilised, sterile

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR