ODN 1826 (synthetisch), TLRgrade™

Lieferant: ENZO LIFE SCIENCES

ALX746002C100
ENZOALX746002C100EA 237 EUR
ENZOALX746002C100
ODN 1826 (synthetisch), TLRgrade™
ODN 1826

TLR9 ligand.


  • Activity and endotoxin tested


CpG oligodeoxynucleotide (Type B) with phosphorothioate backbone. Specific ligand for mouse TLR9 (Toll-like receptor 9).


Sufficient for at least 100 cellular activation assays at 0.1 – 0.5 µg/well when performed in a 200 µl assay volume (i.e. 96 well microtiter plate).


Lieferumfang: Includes 1 vial of double distilled water (endotoxin-free).

Bestellen

Spezifikation Testergebnisse

Source Synthetic
Formulation Lyophilised. Sterile.
Purity Activity and endotoxin tested - TLRgrade™
Endotoxin content <0.002 EU/µg
Quantity 15 nmol (~96µg)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR