ODN 1585 (synthetisch), TLRgrade™

Lieferant: ENZO LIFE SCIENCES

ALX746003C100
ENZOALX746003C100EA 234 EUR
ENZOALX746003C100
ODN 1585 (synthetisch), TLRgrade™
ODN 1585

TLR9 ligand.


CpG oligodeoxynucleotide (Type A) optimised for NK cell activation with mixed phosphodiester/phosphorothioate backbone. Specific ligand for mouse TLR9 (Toll-like receptor 9).


Sufficient for at least 100 cellular activation assays at 0.1 – 0.5 µg/well when performed in a 200 µl assay volume (i.e. 96 well microtiter plate).


Lieferumfang: Includes 1 vial of double distilled water (endotoxin-free).

Bestellen

Spezifikation Testergebnisse

Source Synthetic
Formulation Lyophilised. Sterile.
Purity Activity and endotoxin tested - TLRgrade™
Endotoxin content <0.002 EU/µg
Quantity 14.9 nmol (~99µg)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR