ODN 1668 (synthetisch), TLRgrade™, steril, gefriergetrocknet

Lieferant: ENZO LIFE SCIENCES

ALX746051M001
ENZOALX746051M001EA 841 EUR
ENZOALX746051M001
ODN 1668 (synthetisch), TLRgrade™, steril, gefriergetrocknet
ODN 1668

TLR9 ligand.


CpG oligodeoxynucleotide (Type B) with phosphorothioate backbone. Specific ligand for mouse TLR9 (Toll-like receptor 9).


Lieferumfang: Includes 1 vial of double distilled water (endotoxin-free).

Bestellen

Spezifikation Testergebnisse

Source Synthetic
Formulation Lyophilised. Sterile.
Purity Activity and endotoxin tested - TLRgrade™
Endotoxin content <0.002 EU/µg
Quantity 157.1 nmol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR