ODN 1585 (synthetisch), TLRgrade™

Lieferant: ENZO LIFE SCIENCES

ALX746053M001
ENZOALX746053M001EA 841 EUR
ENZOALX746053M001
ODN 1585 (synthetisch), TLRgrade™
ODN 1585

TLR9 ligand.


CpG oligodeoxynucleotide (Type A) optimised for NK cell activation with mixed phosphodiester/phosphorothioate backbone. Specific ligand for mouse TLR9 (Toll-like receptor 9).


Lieferumfang: Includes 1 vial of double distilled water (endotoxin-free).

Bestellen

Spezifikation Testergebnisse

Source Synthetic
Formulation Lyophilised. Sterile.
Purity Activity and endotoxin tested - TLRgrade™
Endotoxin content <0.002 EU/µg
Quantity 150 nmol (~1000 µg)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR